AC5410537706 Accela ChemBio Inc.

Methyl 1-(2-Methoxyethyl)-2-oxopyrrolidine-4-carboxylate

  • MDL number MFCD18651721
  • Cas Number : 1272755-99-3
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013255&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013255.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart