AC5410537708 Accela ChemBio Inc.

4-Fluoro-1-methyl-4-piperidinemethanol

  • MDL number MFCD09880180
  • Cas Number : 1000341-04-7
  • Molecular Weight : 147.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013297&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013297.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart