AC5410537717 Accela ChemBio Inc.

6-Fluoropyridine-2-carboxaldehyde

  • MDL number MFCD03095149
  • Cas Number : 208110-81-0
  • Molecular Weight : 125.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014043&attname=GoodsCode

Safety Information

signalword Danger
pcodes P261;P280;P305+P351+P338;P304+P340;P405;P501
hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014043.pdf
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart