AC5410537723 Accela ChemBio Inc.

1-(2,3-Difluorophenyl)ethanol

  • MDL number MFCD18911748
  • Cas Number : 1228690-56-9
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014337&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014337.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart