AC5410537730 Accela ChemBio Inc.

Methyl 3-Pyridylacetate Hydrochloride

  • MDL number MFCD19442777
  • Cas Number : 69966-42-3
  • Molecular Weight : 187.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014800&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014800.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart