AC5410537741 Accela ChemBio Inc.

5-(2-Nitrophenyl)isoxazole-3-carboxylic Acid

  • MDL number MFCD17179148
  • Cas Number : 1232028-11-3
  • Molecular Weight : 234.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015159&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015159.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart