AC5410537745 Accela ChemBio Inc.

Methyl 5-(4-Nitrophenyl)isoxazole-3-carboxylate

  • MDL number MFCD02159572
  • Cas Number : 487034-01-5
  • Molecular Weight : 248.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015164&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015164.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart