AC5410537748 Accela ChemBio Inc.

5-(4-Isopropylphenyl)isoxazole-3-carboxylic Acid

  • MDL number MFCD07377124
  • Cas Number : 33282-10-9
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015167&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015167.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart