AC5410537752 Accela ChemBio Inc.

5-(2-Pyridyl)isoxazole-3-carboxylic Acid

  • MDL number MFCD07377113
  • Cas Number : 893638-37-4
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015175&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015175.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart