AC5410537757 Accela ChemBio Inc.

Methyl 5-(3-Cyanophenyl)isoxazole-3-carboxylate

  • MDL number MFCD22192336
  • Cas Number : 956360-06-8
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015182&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015182.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart