AC5410537761 Accela ChemBio Inc.

5-(1-Methyl-2-pyrrolyl)isoxazole-3-carboxylic Acid

  • MDL number MFCD19706940
  • Cas Number : 1326814-81-6
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015187&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015187.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart