AC5410537762 Accela ChemBio Inc.

Methyl 5-(1-Methyl-2-pyrrolyl)isoxazole-3-carboxylate

  • MDL number MFCD22192341
  • Cas Number : 1375064-53-1
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015188&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015188.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart