AC5410537764 Accela ChemBio Inc.

Methyl 5-(2,4-Difluorophenyl)isoxazole-3-carboxylate

  • MDL number MFCD11986431
  • Cas Number : 1105191-49-8
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015191&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015191.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart