AC5410537765 Accela ChemBio Inc.

5-(2,4-Dichloro-5-fluorophenyl)isoxazole-3-carboxylic Acid

  • MDL number MFCD06199330
  • Cas Number : 763109-73-5
  • Molecular Weight : 276.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015192&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015192.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart