AC5410537766 Accela ChemBio Inc.

Ethyl 5-(3-Nitrophenyl)isoxazole-3-carboxylate

  • MDL number MFCD03848619
  • Cas Number : 866040-66-6
  • Molecular Weight : 262.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015193&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015193.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart