AC5410537767 Accela ChemBio Inc.

Ethyl 3-Oxo-3-(2-tetrahydrofuryl)propanoate

  • MDL number MFCD06410652
  • Cas Number : 887411-85-0
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015215&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015215.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart