AC5410537775 Accela ChemBio Inc.

4-tert-Butoxybenzyl Alcohol

  • MDL number MFCD00082599
  • Cas Number : 51503-08-3
  • Molecular Weight : 180.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015586&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015586.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart