AC5410537781 Accela ChemBio Inc.

3-(Cbz-amino)-3-(4-piperidyl)propionic Acid

  • MDL number MFCD03425244
  • Cas Number : 372144-06-4
  • Molecular Weight : 306.35688

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015777&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015777.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart