AC5410537783 Accela ChemBio Inc.

1-Cbz-N-methoxy-N-methyl-4-piperidinecarboxamide

  • MDL number MFCD07370044
  • Cas Number : 148148-48-5
  • Molecular Weight : 306.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015842&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015842.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart