AC5410537787 Accela ChemBio Inc.

2-Amino-2-[3-(trifluoromethyl)phenyl]acetic Acid Hydrochloride

  • MDL number MFCD07371458
  • Cas Number : 1134915-25-5
  • Molecular Weight : 255.6214696

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015957&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015957.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart