AC5410537789 Accela ChemBio Inc.

2-Amino-5-iodobenzonitrile

  • MDL number MFCD07787432
  • Cas Number : 132131-24-9
  • Molecular Weight : 244.03247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016027&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016027.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart