AC5410537794 Accela ChemBio Inc.

Ethyl 3-Amino-1-benzyl-3-pyrrolidinecarboxylate

  • MDL number MFCD06410513
  • Cas Number : 475469-12-6
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016276&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016276.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart