AC5410537795 Accela ChemBio Inc.

Ethyl 4-(4-Pyridyl)-2-thiazolecarboxylate

  • MDL number MFCD06797362
  • Cas Number : 216867-38-8
  • Molecular Weight : 234.2743

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016277&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016277.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart