AC5410537796 Accela ChemBio Inc.

2-(Boc-amino)-2-(3-thiophenyl)acetic Acid

  • MDL number MFCD02683176
  • Cas Number : 40512-57-0
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016296&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016296.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart