AC5410537800 Accela ChemBio Inc.

2-Chloro-6,7-dimethoxyquinazoline

  • MDL number MFCD06249130
  • Cas Number : 94644-47-0
  • Molecular Weight : 224.64366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016558&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016558.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart