AC5410537824 Accela ChemBio Inc.

1-(Cyclopropylmethyl)-5-oxopyrrolidine-3-carboxylic Acid

  • MDL number MFCD07379775
  • Cas Number : 845546-16-9
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018542&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018542.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart