AC5410537826 Accela ChemBio Inc.

1-Ethyl-7-methoxy-2-tetralone

  • MDL number MFCD21364427
  • Cas Number : 3238-50-4
  • Molecular Weight : 204.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018568&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018568.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart