AC5410537831 Accela ChemBio Inc.

4-[(Methylamino)methyl]phenol

  • MDL number MFCD09046544
  • Cas Number : 78507-19-4
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018667&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY018667.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart