AC5410537832 Accela ChemBio Inc.

4-[(Ethylamino)methyl]phenol

  • MDL number MFCD09929168
  • Cas Number : 45966-19-6
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018668&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY018668.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart