AC5410537835 Accela ChemBio Inc.

Ethyl 1-Boc-3-methylpiperidine-3-carboxylate

  • MDL number MFCD11559117
  • Cas Number : 278789-43-8
  • Molecular Weight : 271.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018677&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018677.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart