AC5410537841 Accela ChemBio Inc.

4-Chloro-2-[(methylamino)methyl]phenol

  • MDL number MFCD09044328
  • Cas Number : 38926-77-1
  • Molecular Weight : 171.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018708&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY018708.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart