AC5410537845 Accela ChemBio Inc.

2-Amino-3-bromo-5-hydroxypyridine

  • MDL number MFCD09835227
  • Cas Number : 186593-53-3
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-5-bromo-3-pyridinol
Inchi Code 1S/C5H5BrN2O/c6-4-1-3(9)2-8-5(4)7/h1-2,9H,(H2,7,8)
Inchi Key BVZMUNKWVZDBOL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018764&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018764.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart