AC5410537848 Accela ChemBio Inc.

N-(2-Chloroethyl)methanesulfonamide

  • MDL number MFCD00129996
  • Cas Number : 21896-59-3
  • Molecular Weight : 157.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018853&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018853.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart