AC5410537895 Accela ChemBio Inc.

2,2-Dimethyltetrahydropyran-4-carbaldehyde

  • MDL number MFCD01822189
  • Cas Number : 34941-21-4
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Colorless to yellow liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022720&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022720.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart