AC5410537941 Accela ChemBio Inc.

5-Phenyluracil

  • MDL number MFCD06801865
  • Cas Number : 15761-83-8
  • Molecular Weight : 188.18272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024184&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024184.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart