AC5410537950 Accela ChemBio Inc.

4,5-Dibromo-6,7-dihydro-1H-indeno[5,4-b]furan-8(2H)-one

  • MDL number MFCD09955084
  • Cas Number : 196597-77-0
  • Molecular Weight : 331.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024366&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024366.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart