AC5410537951 Accela ChemBio Inc.

5-Amino-2-(methylthio)pyrimidine

  • MDL number MFCD10697095
  • Cas Number : 42382-46-7
  • Molecular Weight : 141.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024373&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024373.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart