AC5410537956 Accela ChemBio Inc.

5-Amino-2-methyl-2-pentanol

  • MDL number MFCD19210993
  • Cas Number : 108262-66-4
  • Molecular Weight : 117.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024445&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024445.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart