AC5410537961 Accela ChemBio Inc.

N-Ethyl-2-(1-piperidyl)ethanamine

  • MDL number MFCD08361609
  • Cas Number : 32776-21-9
  • Molecular Weight : 156.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024484&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY024484.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart