AC5410537963 Accela ChemBio Inc.

N-Ethyl-2-(1-imidazolyl)ethanamine

  • MDL number MFCD14619072
  • Cas Number : 1211472-59-1
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024490&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY024490.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart