AC5410537969 Accela ChemBio Inc.

Ethyl (1R,2R)-2-Aminocyclopentanecarboxylate

  • MDL number MFCD20719057
  • Cas Number : 1609100-30-2
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (1R,2R)-2-aminocyclopentanecarboxylate
Inchi Code 1S/C8H15NO2/c1-2-11-8(10)6-4-3-5-7(6)9/h6-7H,2-5,9H2,1H3/t6-,7-/m1/s1
Inchi Key AGCJYTRMZBPEEO-RNFRBKRXSA-N

Description

Purity 95%
Storage temp. -20?
Physical Form Colorless to yellow liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024586&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024586.pdf
hcodes No data known
Availability details for
Quantity
  • Availability
  • Add to Cart