AC5410537983 Accela ChemBio Inc.

N-Boc-N-(3-piperidyl)cyclopropylamine

  • MDL number MFCD21098142
  • Cas Number : 250275-24-2
  • Molecular Weight : 240.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024720&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY024720.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart