AC5410537987 Accela ChemBio Inc.

3-[2-(Dimethylamino)vinyl]-4-nitropyridine 1-Oxide

  • MDL number MFCD12405620
  • Cas Number : 104118-88-9
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Light purple to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024746&attname=GoodsCode

Safety Information

signalword Warning
MSDS http://pdf.accelachem.com/cn/msds/SY024746.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart