AC5410537996 Accela ChemBio Inc.

N-Ethyl-2-(1-pyrazolyl)acetamide

  • MDL number MFCD18840072
  • Cas Number : 1205076-85-2
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024906&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024906.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart