AC5410538001 Accela ChemBio Inc.

2,4-Dibenzyloxybenzyl Alcohol

  • MDL number MFCD08456666
  • Cas Number : 33617-58-2
  • Molecular Weight : 320.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024921&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024921.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart