AC5410538004 Accela ChemBio Inc.

2-Chloro-4-fluorobenzotrifluoride

  • MDL number MFCD03412197
  • Cas Number : 94444-58-3
  • Molecular Weight : 198.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024935&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024935.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart