AC5410538005 Accela ChemBio Inc.

4-Bromo-1-(3-hydroxypropyl)pyrazole

  • MDL number MFCD06804869
  • Cas Number : 925180-06-9
  • Molecular Weight : 205.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024944&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024944.pdf
pictograms No data known
Availability details for
Quantity
  • Availability
  • Add to Cart