AC5410538006 Accela ChemBio Inc.

3-[Benzyl(methyl)amino]propanoic Acid

  • MDL number MFCD09717730
  • Cas Number : 149692-49-9
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024957&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024957.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart