AC5410538020 Accela ChemBio Inc.

2',4'-Bis(benzyloxy)acetophenone

  • MDL number MFCD00101153
  • Cas Number : 22877-01-6
  • Molecular Weight : 332.3924

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025208&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025208.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart