AC5410538022 Accela ChemBio Inc.

4,4'-Diamino-2,2'-difluorobiphenyl

  • MDL number MFCD19302620
  • Cas Number : 316-64-3
  • Molecular Weight : 220.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025242&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025242.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart